2-Bromobenzimidazole 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-1H-benzimidazole

OR23118-1G OR23118-5G
APOSOR23118-1GEA 37.7 EUR
APOSOR23118-1G APOSOR23118-5G
2-Bromobenzimidazole 99%
2-Bromobenzimidazole
Formula: C₇H₅BrN₂
Melting Pt: 200…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00160009
CAS Number: 54624-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR