5-Phenyl-2-thiophenecarbaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Phenylthiophene-2-carbaldehyde, 5-Phenylthiophene-2-carboxaldehyde
OR23148-1G
OR23148-5G
APOSOR23148-1GEA
120
EUR
APOSOR23148-1G
APOSOR23148-5G
5-Phenyl-2-thiophenecarbaldehyde
5-Phenyl-2-thiophenecarbaldehyde
Formula:
C₁₁H₈OS MW: 188,25 g/mol Melting Pt: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD01151752 CAS Number: 19163-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...