Benzo[b]thiophene-3-carboxaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Benzo[b]thiophene-3-carbaldehyde
OR23174-100G
OR23174-25G
OR23174-5G
APOSOR23174-100GEA
503
EUR
APOSOR23174-100G
APOSOR23174-25G
APOSOR23174-5G
Benzo[b]thiophene-3-carboxaldehyde 95%
Benzo[b]thiophene-3-carboxaldehyde
Formula:
C₉H₆OS MW: 162,2083 g/mol Boiling Pt: 166 °C/20mmHg Melting Pt: 53…57 °C Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052376 CAS Number: 5381-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...