4-Benzyl-2-morpholinecarboxylic acid hydrochloride
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
4-Benzylmorpholine-2-carboxylic acid hydrochloride
OR23182-1G
OR23182-250MG
OR23182-5G
APOSOR23182-1GEA
29
EUR
APOSOR23182-1G
APOSOR23182-250MG
APOSOR23182-5G
4-Benzyl-2-morpholinecarboxylic acid hydrochloride
4-Benzyl-2-morpholinecarboxylic acid hydrochloride
Formula:
C₁₂H₁₆ClNO₃ MW: 257,71 g/mol Melting Pt: 244…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682031 CAS Number: 135072-15-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...