4-Benzylmorpholine-2-carbonylchloride hydrochloride
Supplier: Apollo Scientific
|
Warning
|
OR23183-1G
OR23183-100MG
OR23183-250MG
OR23183-500MG
APOSOR23183-1GEA
589
EUR
APOSOR23183-1G
APOSOR23183-100MG
APOSOR23183-250MG
APOSOR23183-500MG
4-Benzylmorpholine-2-carbonylchloride hydrochloride
4-Benzylmorpholine-2-carbonylchloride hydrochloride
Formula:
C₁₂H₁₅Cl₂NO₂ MW: 276.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407323 CAS Number: 135072-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...