α-Bromo-o-toluonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Cyanobenzyl bromide, 2-(Bromomethyl)benzonitrile, o-(Bromomethyl)benzonitrile
OR2319-100G
OR2319-25G
OR2319-500G
APOSOR2319-100GEA
40.6
EUR
APOSOR2319-100G
APOSOR2319-25G
APOSOR2319-500G
α-Bromo-o-toluonitrile 98%
α-Bromo-o-toluonitrile
Formula:
C₈H₆BrN MW: 196,05 g/mol Boiling Pt: 111 °C/1,5mmHg Melting Pt: 72…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001794 CAS Number: 22115-41-9 UN: 3261 ADR: 8,II |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...