2-Thienyl isocyanate

Supplier: Apollo Scientific
Warning

OR23197-1G OR23197-5G OR23197-100MG OR23197-250MG OR23197-500MG
APOSOR23197-1GEA 215 EUR
APOSOR23197-1G APOSOR23197-5G APOSOR23197-100MG APOSOR23197-250MG APOSOR23197-500MG
2-Thienyl isocyanate
2-Thienyl isocyanate

  • May form small amount of sediment.

Formula: C₅H₃NOS
MW: 125,15 g/mol
Boiling Pt: 36…38 °C/1mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD02677700
CAS Number: 2048-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR