tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,9-Diazaspiro[5.5]undecane, N9-BOC protected
OR2329-1G
OR2329-5G
APOSOR2329-1GEA
1650
EUR
APOSOR2329-1G
APOSOR2329-5G
tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate
Formula:
C₁₄H₂₆N₂O₂ MW: 254,38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09998745 CAS Number: 1031927-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...