(4-Imidazol-1-ylphenyl)methanol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(1H-Imidazol-1-yl)benzyl alcohol, (4-Imidazol-1-yl-phenyl)methanol
OR23300-1G
APOSOR23300-1GEA
21.9
EUR
APOSOR23300-1G
(4-Imidazol-1-ylphenyl)methanol 95%
(4-Imidazol-1-ylphenyl)methanol
Formula:
C₁₀H₁₀N₂O MW: 174,2000 g/mol Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682059 CAS Number: 86718-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...