3-(1-Pyrrolyl)-2-thiophenemethanol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-(1H-Pyrrol-1-yl)thiophen-2-yl)methanol
OR23357-1G
OR23357-250MG
OR23357-5G
APOSOR23357-1GEA
58
EUR
APOSOR23357-1G
APOSOR23357-250MG
APOSOR23357-5G
3-(1-Pyrrolyl)-2-thiophenemethanol
3-(1-Pyrrolyl)-2-thiophenemethanol
Formula:
C₉H₉NOS MW: 179,24 g/mol Boiling Pt: 104…108 °C/1,5mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00052578 CAS Number: 107073-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...