[4-(1H-Pyrrol-1-yl)phenyl]methanol

Supplier: Apollo Scientific

Synonyms: 4-(1H-Pyrrol-1-yl)benzyl alcohol

OR23400-1G OR23400-250MG OR23400-5G OR23400-500MG OR23400-10G OR23400-25G
APOSOR23400-1GEA 119 EUR
APOSOR23400-1G APOSOR23400-250MG APOSOR23400-5G APOSOR23400-500MG APOSOR23400-10G APOSOR23400-25G
[4-(1H-Pyrrol-1-yl)phenyl]methanol
[4-(1H-Pyrrol-1-yl)phenyl]methanol
Formula: C₁₁H₁₁NO
MW: 173,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03659708
CAS Number: 143426-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR