4-Chloro-2-(methylthio)-1-nitrobenzene 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Chloro-2-nitrothioanisole
OR23642-1G
OR23642-250MG
APOSOR23642-1GEA
493
EUR
APOSOR23642-1G
APOSOR23642-250MG
4-Chloro-2-(methylthio)-1-nitrobenzene 95%
4-Chloro-2-(methylthio)-1-nitrobenzene
Formula:
C₇H₆ClNO₂S MW: 203,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052209 CAS Number: 70019-41-9 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...