3-Oxo-3-(2-thienyl)propionitrile
Supplier: Apollo Scientific
|
Warning
|
OR23646-10G
OR23646-1G
OR23646-5G
APOSOR23646-10GEA
105
EUR
APOSOR23646-10G
APOSOR23646-1G
APOSOR23646-5G
3-Oxo-3-(2-thienyl)propionitrile
3-Oxo-3-(2-thienyl)propionitrile
Formula:
C₇H₅NOS MW: 151,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052586 CAS Number: 33898-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...