Ethyl-3,5-dimethylisoxazole-4-carboxylate

Supplier: Apollo Scientific
Warning

OR23656-1G OR23656-5G
APOSOR23656-1GEA 21.9 EUR
APOSOR23656-1G APOSOR23656-5G
Ethyl-3,5-dimethylisoxazole-4-carboxylate
Ethyl-3,5-dimethylisoxazole-4-carboxylate
Formula: C₈H₁₁NO₃
MW: 169,18 g/mol
Boiling Pt: 87…89 °C/2mmHg
Density: 1,11 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00052557
CAS Number: 17147-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR