1-tert-Butyl-1H-pyrrole

Supplier: Apollo Scientific

OR23801-1G OR23801-250MG OR23801-500MG OR23801-5G
APOSOR23801-1GEA 258 EUR
APOSOR23801-1G APOSOR23801-250MG APOSOR23801-500MG APOSOR23801-5G
1-tert-Butyl-1H-pyrrole
1-tert-Butyl-1H-pyrrole
Formula: C₈H₁₃N
MW: 123.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015558
CAS Number: 24764-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR