4-Phenylbutyric acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Phenylbutanoic acid
OR2390-100G
OR2390-25G
OR2390-500G
APOSOR2390-100GEA
94.4
EUR
APOSOR2390-100G
APOSOR2390-25G
APOSOR2390-500G
4-Phenylbutyric acid 98%
4-Phenylbutyric acid
Formula:
C₁₀H₁₂O₂ MW: 164,2 g/mol Boiling Pt: 165 °C/10mmHg Melting Pt: 49…53 °C Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004403 CAS Number: 1821-12-1 EINECS: 217-341-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...