4-Acetoxyindole 98%
Supplier: Apollo Scientific
|
Warning
|
OR2401-100G
OR2401-5G
OR2401-25G
APOSOR2401-100GEA
729
EUR
APOSOR2401-100G
APOSOR2401-5G
APOSOR2401-25G
4-Acetoxyindole 98%
4-Acetoxyindole
Formula:
C₁₀H₉NO₂ MW: 175,19 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010678 CAS Number: 5585-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...