5-Nitrothiophene-3-carbaldehyde 97%
Supplier: Apollo Scientific
|
Warning
|
OR24041-1G
APOSOR24041-1GEA
95.7
EUR
APOSOR24041-1G
5-Nitrothiophene-3-carbaldehyde 97%
5-Nitrothiophene-3-carbaldehyde
Formula:
C₅H₃NO₃S MW: 157,15 g/mol Melting Pt: 79…81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052581 CAS Number: 75428-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...