3-[(3-Pyridinylmethyl)amino]propanenitrile
Supplier: Apollo Scientific
|
Warning
|
OR24149-10G
OR24149-1G
OR24149-5G
APOSOR24149-10GEA
169
EUR
APOSOR24149-10G
APOSOR24149-1G
APOSOR24149-5G
3-[(3-Pyridinylmethyl)amino]propanenitrile
3-[(3-Pyridinylmethyl)amino]propanenitrile
Formula:
C₉H₁₁N₃ Boiling Pt: 241 °C/50mmHg Density: 1,078 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047455 CAS Number: 33611-48-2 EINECS: 251-596-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...