Ethyl-2-amino-4-methylthiophene-3-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-amino-4-methylthiophene-3-carboxylate

OR24230-10G OR24230-25G
APOSOR24230-10GEA 65.5 EUR
APOSOR24230-10G APOSOR24230-25G
Ethyl-2-amino-4-methylthiophene-3-carboxylate
Ethyl 2-amino-4-methylthiophene-3-carboxylate
Formula: C₈H₁₁NO₂S
MW: 185,25 g/mol
Melting Pt: 79…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051669
CAS Number: 43088-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR