1-(4-Chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR24329-10G OR24329-1G
APOSOR24329-10GEA 379 EUR
APOSOR24329-10G APOSOR24329-1G
1-(4-Chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
1-(4-Chlorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid
Formula: C₁₃H₁₃ClN₂O₂
Melting Pt: 116…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00068141
CAS Number: 175137-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR