1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone

Supplier: Apollo Scientific
Warning

OR24363-1G OR24363-500MG OR24363-5G
APOSOR24363-1GEA 325 EUR
APOSOR24363-1G APOSOR24363-500MG APOSOR24363-5G
1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone
1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone
Formula: C₉H₆ClN₃O₃
MW: 239,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00099555
CAS Number: 68159-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR