2-Bromo-9-fluorenone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-9H-fluoren-9-one
OR2491-1G
OR2491-100G
OR2491-500G
APOSOR2491-1GEA
21.9
EUR
APOSOR2491-1G
APOSOR2491-100G
APOSOR2491-500G
2-Bromo-9-fluorenone
2-Bromo-9-fluorenone
Formula:
C₁₃H₇BrO MW: 259,1 g/mol Melting Pt: 146…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010789 CAS Number: 3096-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...