4-Nitro-1-naphthylamine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Nitronaphthalen-1-amine, 1-Amino-4-nitronaphthalene

OR24922-25G OR24922-5G OR24922-10G
APOSOR24922-25GEA 116 EUR
APOSOR24922-25G APOSOR24922-5G APOSOR24922-10G
4-Nitro-1-naphthylamine
4-Nitro-1-naphthylamine
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00004026
CAS Number: 776-34-1
EINECS: 212-277-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR