1-(2-Pyridinyl)-1-ethanone oxime

Supplier: Apollo Scientific
Warning

OR25030-1G OR25030-5G
APOSOR25030-1GEA 87.3 EUR
APOSOR25030-1G APOSOR25030-5G
1-(2-Pyridinyl)-1-ethanone oxime
1-(2-Pyridinyl)-1-ethanone oxime
Formula: C₇H₈N₂O
MW: 136,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00499500
CAS Number: 1758-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR