Di-2-thienyl sulphide 97%

Supplier: Apollo Scientific

Synonyms: 2-(2-thienylthio)thiophene

OR25442-1G OR25442-5G
APOSOR25442-1GEA 0 EUR
APOSOR25442-1G APOSOR25442-5G
Di-2-thienyl sulphide 97%
Di-2-thienyl sulphide
Formula: C₈H₆S₃
MW: 198,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052047
CAS Number: 3988-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR