3,4,5-Trichloropyridine 98%

Supplier: Apollo Scientific
Warning

OR2545-100G OR2545-25G
APOSOR2545-100GEA 55.3 EUR
APOSOR2545-100G APOSOR2545-25G
3,4,5-Trichloropyridine 98%
3,4,5-Trichloropyridine
Formula: C₅H₂Cl₃N
MW: 182,44 g/mol
Boiling Pt: 215 °C
Melting Pt: 75…77 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051685
CAS Number: 33216-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR