3-(4,6-Dimethylpyrimidin-2-oxy)benzoic acid 95%

Supplier: Apollo Scientific
Warning

OR2554-1G OR2554-250MG
APOSOR2554-1GEA 546 EUR
APOSOR2554-1G APOSOR2554-250MG
3-(4,6-Dimethylpyrimidin-2-oxy)benzoic acid 95%
3-(4,6-Dimethylpyrimidin-2-oxy)benzoic acid
Formula: C₁₃H₁₂N₂O₃
MW: 244,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09262519
CAS Number: 927981-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR