2-Amino-5-nitro-6-picoline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine
OR2556-25G
OR2556-5G
OR2556-100G
APOSOR2556-25GEA
16.1
EUR
APOSOR2556-25G
APOSOR2556-5G
APOSOR2556-100G
2-Amino-5-nitro-6-picoline
2-Amino-5-nitro-6-picoline
Formula:
C₆H₇N₃O₂ MW: 153,14 g/mol Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053582 CAS Number: 22280-62-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...