4-Methylpyridazine 97%

Supplier: Apollo Scientific
Warning

OR25618-25G
APOSOR25618-25GEA 208 EUR
APOSOR25618-25G
4-Methylpyridazine 97%
4-Methylpyridazine
Formula: C₅H₆N₂
MW: 94,12 g/mol
Boiling Pt: 98…100 °C/11mmHg
Density: 1,06 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00011594
CAS Number: 1120-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR