Ethyl-2-Amino-4-phenylthiazole-5-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-4-phenylthiazole-5-carboxylic Acid Ethyl Ester, Ethyl 2-amino-4-phenylthiazole-5-carboxylate

OR2569-1G OR2569-5G
APOSOR2569-1GEA 79.9 EUR
APOSOR2569-1G APOSOR2569-5G
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate
Formula: C₁₂H₁₂N₂O₂S
Melting Pt: 167…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00022452
CAS Number: 64399-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR