Isopropoxybenzene
Supplier: Apollo Scientific
OR25710-100G
OR25710-25G
OR25710-5G
APOSOR25710-100GEA
188
EUR
APOSOR25710-100G
APOSOR25710-25G
APOSOR25710-5G
Isopropoxybenzene
Isopropoxybenzene
Formula:
C₉H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD00048288 CAS Number: 2741-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...