7-Aminoenanthic acid 98%

Supplier: Apollo Scientific

OR2575-100G OR2575-25G
APOSOR2575-100GEA 699 EUR
APOSOR2575-100G APOSOR2575-25G
7-Aminoenanthic acid 98%
7-Aminoenanthic acid
Formula: C₇H₁₅NO₂
Melting Pt: 192…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00008242
CAS Number: 929-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR