6-(4-Methylpiperazin-1-yl)nicotinic acid

Supplier: Apollo Scientific
Warning

OR2591-1G OR2591-250MG
APOSOR2591-1GEA 301 EUR
APOSOR2591-1G APOSOR2591-250MG
6-(4-Methylpiperazin-1-yl)nicotinic acid
6-(4-Methylpiperazin-1-yl)nicotinic acid
Formula: C₁₁H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07376841
CAS Number: 132521-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR