5-Methylthiazole-2-carboxylic acid 98%

Supplier: Apollo Scientific
Warning

OR2597-1G OR2597-250MG OR2597-5G
APOSOR2597-1GEA 61.1 EUR
APOSOR2597-1G APOSOR2597-250MG APOSOR2597-5G
5-Methylthiazole-2-carboxylic acid 98%
5-Methylthiazole-2-carboxylic acid
Formula: C5H5NO2S
MW: 143,16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08060736
CAS Number: 61291-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR