2-(4-Methoxyphenyl)propan-2-ol
Supplier: Apollo Scientific
|
Warning
|
OR2602-1G
OR2602-250MG
OR2602-500MG
OR2602-2.5G
APOSOR2602-1GEA
364
EUR
APOSOR2602-1G
APOSOR2602-250MG
APOSOR2602-500MG
APOSOR2602-2.5G
2-(4-Methoxyphenyl)propan-2-ol
2-(4-Methoxyphenyl)propan-2-ol
Formula:
C₁₀H₁₄O₂ Boiling Pt: 100…101 °C/0,37mmHg Melting Pt: 37…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD02066307 CAS Number: 7428-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...