1,3-Dimethyl-5-pyrazolone 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one, 1,3-Dimethyl-1H-pyrazol-5(4H)-one
OR26105-100G
APOSOR26105-100GEA
0
EUR
APOSOR26105-100G
1,3-Dimethyl-5-pyrazolone 95%
1,3-Dimethyl-5-pyrazolone
Formula:
C₅H₈N₂O MW: 112,13 g/mol Melting Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067008 CAS Number: 2749-59-9 EINECS: 220-389-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...