2,4-Diamino-6-methyl-1,3,5-triazine 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,4-Diamino-6-methyl-1,3,5-triazine 6-Methyl-1,3,5-triazine-2,4-diamine, 2,4-Diamino-6-methyl-s-triazine, 6-Methyl-1,3,5-triazine-2,4-diamine, Acetoguanamine, 6-Methyl-1,3,5-triazine-2,4-diyldiamine
OR26241-100G
OR26241-500G
APOSOR26241-100GEA
43.5
EUR
APOSOR26241-100G
APOSOR26241-500G
2,4-Diamino-6-methyl-1,3,5-triazine 96%
2,4-Diamino-6-methyl-1,3,5-triazine
Formula:
C₄H₇N₅ MW: 125,13 g/mol Melting Pt: 274…276 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023192 CAS Number: 542-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...