(4-Methyl-1,2,3-thiadiazol-5-yl)methanol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(Hydroxymethyl)-4-methyl-1,2,3-thiadiazole
OR2642-10G
OR2642-1G
OR2642-5G
APOSOR2642-10GEA
582
EUR
APOSOR2642-10G
APOSOR2642-1G
APOSOR2642-5G
(4-Methyl-1,2,3-thiadiazol-5-yl)methanol 95%
(4-Methyl-1,2,3-thiadiazol-5-yl)methanol
Formula:
C₄H₆N₂OS MW: 130,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371384 CAS Number: 163008-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...