4-Amino-3-methoxyphenylboronic acid pinacol ester 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-3-methoxyphenylboronic acid, pinacol ester, 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

OR2652-1G OR2652-5G
APOSOR2652-1GEA 71.2 EUR
APOSOR2652-1G APOSOR2652-5G
4-Amino-3-methoxyphenylboronic acid pinacol ester 98%
4-Amino-3-methoxyphenylboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249,12 g/mol
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD06795657
CAS Number: 461699-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR