N-Methylhydrazinecarboximidamide hydroiodide 95%

Supplier: Apollo Scientific
Warning

OR26561-1G OR26561-5G OR26561-10G
APOSOR26561-1GEA 239 EUR
APOSOR26561-1G APOSOR26561-5G APOSOR26561-10G
N-Methylhydrazinecarboximidamide hydroiodide 95%
N-Methylhydrazinecarboximidamide hydroiodide
Formula: C₂H₉IN₄
MW: 216,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179313
CAS Number: 33398-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR