5-Methoxy-2-nitrophenylamine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-4-methoxy-1-nitrobenzene, 5-Methoxy-2-nitroaniline, 3-Amino-4-nitroanisole
OR26572-25G
OR26572-100G
APOSOR26572-25GEA
109
EUR
APOSOR26572-25G
APOSOR26572-100G
5-Methoxy-2-nitrophenylamine
5-Methoxy-2-nitrophenylamine
Formula:
C7H8N2O3 MW: 168,15 g/mol Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00179573 CAS Number: 16133-49-6 EINECS: 240-293-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...