4-Nitrochalcone

Supplier: Apollo Scientific
Warning

Synonyms: 4-Nitrobenzylidenacetophenone

OR26594-25G OR26594-5G
APOSOR26594-25GEA 92.8 EUR
APOSOR26594-25G APOSOR26594-5G
4-Nitrochalcone
4-Nitrochalcone
Formula: C₁₅H₁₁NO₃
MW: 253,26 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00007382
CAS Number: 1222-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR