Ethyl-4-amino-3,5-diiodobenzoate

Supplier: Apollo Scientific
Warning

OR26788-5G OR26788-10G OR26788-1G
APOSOR26788-5GEA 92.8 EUR
APOSOR26788-5G APOSOR26788-10G APOSOR26788-1G
Ethyl-4-amino-3,5-diiodobenzoate
Ethyl 4-amino-3,5-diiodobenzoate
Formula: C₉H₉I₂NO₂
MW: 416,982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017092
CAS Number: 5400-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR