3,4-Pyridinedicarboxylic acid anhydride

Supplier: Apollo Scientific
Warning

Synonyms: Furo[3,4-c]pyridine-1,3-dione

OR26823-25G OR26823-5G OR26823-1G OR26823-10G OR26823-50G
APOSOR26823-25GEA 132 EUR
APOSOR26823-25G APOSOR26823-5G APOSOR26823-1G APOSOR26823-10G APOSOR26823-50G
3,4-Pyridinedicarboxylic acid anhydride
3,4-Pyridinedicarboxylic acid anhydride
Formula: C₇H₃NO₃
MW: 149,11 g/mol
Boiling Pt: 112 °C/3mmHg
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00010680
CAS Number: 4664-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR