4-Amino-3,5-dichloropyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine
OR27150-25G
OR27150-100G
APOSOR27150-25GEA
59.7
EUR
APOSOR27150-25G
APOSOR27150-100G
4-Amino-3,5-dichloropyridine
4-Amino-3,5-dichloropyridine
Formula:
C₅H₄Cl₂N₂ MW: 163,01 g/mol Melting Pt: 159…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00125023 CAS Number: 22889-78-7 EINECS: 245-304-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...