2,6-Dichloroisonicotinonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Cyano-2,6-dichloropyridine
OR27160-1G
OR27160-500MG
OR27160-5G
APOSOR27160-1GEA
51
EUR
APOSOR27160-1G
APOSOR27160-500MG
APOSOR27160-5G
2,6-Dichloroisonicotinonitrile
2,6-Dichloroisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula:
C₆H₂Cl₂N₂ MW: 172,9995 g/mol Melting Pt: 96…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173784 CAS Number: 32710-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...