2-Chloro-6-methoxyisonicotinic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-6-methoxypyridine-4-carboxylic acid

OR27363-1G OR27363-5G OR27363-10G
APOSOR27363-1GEA 31.9 EUR
APOSOR27363-1G APOSOR27363-5G APOSOR27363-10G
2-Chloro-6-methoxyisonicotinic acid
2-Chloro-6-methoxyisonicotinic acid
Formula: C₇H₆ClNO₃
MW: 187,58 g/mol
Boiling Pt: 192…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00173928
CAS Number: 15855-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR