6-Chloronicotinamide
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloropyridine-3-carboxamide
OR27410-25G
OR27410-5G
APOSOR27410-25GEA
187
EUR
APOSOR27410-25G
APOSOR27410-5G
6-Chloronicotinamide
6-Chloronicotinamide
Formula:
C₆H₅ClN₂O MW: 156,57 g/mol Melting Pt: 214…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006242 CAS Number: 6271-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...