Imidazole-4-carboxaldehyde 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1H-Imidazole-5-carboxaldehyde, 4-Formylimidazole, Imidazole-4-carbaldehyde

OR2743-25G OR2743-1G OR2743-5G
APOSOR2743-25GEA 66.7 EUR
APOSOR2743-25G APOSOR2743-1G APOSOR2743-5G
Imidazole-4-carboxaldehyde 98%
Imidazole-4-carboxaldehyde
Formula: C₄H₄N₂O
Melting Pt: 174…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00173726
CAS Number: 3034-50-2
EINECS: 221-227-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR